Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098616.2 | NP_001092086.1 | 3 | 601 | ||
NM_001297717.1 | NP_001284646.1 | 3 | 601 | |||
NM_001297718.1 | NP_001284647.1 | 3 | 524 | |||
NM_001297720.1 | NP_001284649.1 | 3 | 601 | |||
NM_001297721.1 | NP_001284650.1 | 2 | 499 | |||
NM_001297723.1 | NP_001284652.1 | 3 | 601 | |||
NM_015449.3 | NP_056264.1 | 2 | 499 | |||
NM_138740.3 | NP_620077.1 | 2 | 499 | |||
GenBank mRNA | AF077036.1 | - | 2 | 319 | ||
AF109185.1 | - | 3 | 292 | |||
AF529365.1 | - | 3 | 211 | |||
AJ243663.1 | - | 2 | 321 | |||
AJ243664.1 | - | 2 | 267 | |||
AJ243665.1 | - | 2 | 104 | |||
AK290583.1 | - | 3 | 373 | |||
AL117463.1 | - | 1 | 65 | |||
AL540989.3 | - | 3 | 388 | |||
BC000152.2 | - | 3 | 410 | 2901006 | ||
BC001377.2 | - | 2 | 267 | 3051048 | ||
BC006514.1 | - | 2 | 336 | 2822327 | ||
BC008306.1 | - | 3 | 337 | 3502049 | ||
BC014391.1 | - | 2 | 235 | 3544590 | ||
BC088353.1 | - | 3 | 421 | 6164281 | ||
BG478941.1 | - | 3 | 331 | |||
CD247026.1 | - | 2 | 344 | |||
CR457131.1 | - | 2 | 109 | |||
DQ908899.1 | - | 2 | 109 | |||
DR007097.1 | - | 2 | 381 |