Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015450.2 | NP_056265.2 | 7, 8 | 857 | ||
NR_003102.1 | 7, 8 | 857 | ||||
NR_003103.1 | 7, 8 | 857 | ||||
NR_003104.1 | 7, 8 | 857 | ||||
XM_006715917.3 | XP_006715980.1 | 7, 8 | 847 | |||
XM_011516006.1 | XP_011514308.1 | 8, 9 | 969 | |||
XM_011516007.1 | XP_011514309.1 | 8, 9 | 968 | |||
XM_017011941.1 | XP_016867430.1 | 7, 8 | 855 | |||
GenBank mRNA | AK001230.1 | - | 3, 4 | 279 | ||
AK001935.1 | - | 4, 5 | 386 | |||
AK022580.1 | - | 7, 8 | 694 | |||
AK302263.1 | - | 7, 8 | 1012 | |||
AY823520.1 | - | 3, 4 | 267 | |||
AY823521.1 | - | 3, 4 | 267 | |||
AY823522.1 | - | 3, 4 | 267 | |||
AY823524.1 | - | 3, 4 | 267 | |||
BC002923.2 | - | 4, 5 | 313 | 3955573 |