Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168278.1 | NP_001161750.1 | 4 | 736 | ||
NM_001168280.1 | NP_001161752.1 | 3 | 683 | |||
NM_015472.4 | NP_056287.1 | 3 | 819 | |||
XM_017006121.1 | XP_016861610.1 | 6 | 929 | |||
XM_017006122.1 | XP_016861611.1 | 4 | 728 | |||
GenBank mRNA | AJ299431.1 | - | 3 | 654 | ||
AK022036.1 | - | 4 | 736 | |||
AK127620.1 | - | 1 | 682 | |||
AL050107.1 | - | 1 | 43 | |||
AL833852.1 | - | 2 | 473 | |||
BC014052.2 | - | 3 | 713 | 3635491 |