Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271949.1 | NP_001258878.1 | 7 | 1384 | ||
NM_001271950.1 | NP_001258879.1 | 6 | 1360 | |||
NM_015476.3 | NP_056291.2 | 7 | 1489 | |||
XM_017025699.1 | XP_016881188.1 | 8 | 1735 | |||
GenBank mRNA | AB073382.1 | - | 6 | 1130 | ||
AK000543.1 | - | 7 | 1062 | |||
AK295817.1 | - | 7 | 1131 | |||
AL050225.1 | - | 6 | 948 | |||
BC015178.1 | - | 7 | 1250 | 3905315 | ||
BC087845.1 | - | 7 | 1468 | 30390077 |