Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015496.4 | NP_056311.2 | 9 | 2311 | ||
NM_183009.2 | NP_892121.1 | 9 | 2311 | |||
XM_017013306.1 | XP_016868795.1 | 8 | 2091 | |||
XM_017013307.1 | XP_016868796.1 | 8 | 2134 | |||
XM_017013308.1 | XP_016868797.1 | 7 | 2124 | |||
GenBank mRNA | AB037850.1 | - | 9 | 2235 | ||
AF116724.1 | - | 9 | 2235 | |||
AK022906.1 | - | 1 | 82 | |||
AL832487.1 | - | 9 | 2252 | |||
BC003701.1 | - | 2 | 399 | |||
BC069239.1 | - | 2 | 376 | |||
BC112288.1 | - | 9 | 2323 | 8327756 | ||
BC113380.1 | - | 9 | 2311 | 8322432 |