Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282433.1 | NP_001269362.1 | 4 | 829 | ||
NM_015515.4 | NP_056330.3 | 5 | 1357 | |||
XM_005257200.4 | XP_005257257.1 | 4 | 410 | |||
XM_011524595.1 | XP_011522897.1 | 4 | 400 | |||
GenBank mRNA | AF102848.1 | - | 4 | 785 | ||
AK002047.1 | - | 4 | 769 | |||
AK225866.1 | - | 4 | 1140 | |||
AK289449.1 | - | 4 | 852 | |||
AK302109.1 | - | 4 | 638 | |||
AK310880.1 | - | 4 | 941 | |||
AK314604.1 | - | 4 | 569 | |||
AL117538.1 | - | 3 | 613 | |||
BC028356.2 | - | 5 | 1137 | 4838347 |