Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015527.3 | NP_056342.3 | 3, 4 | 1179 | ||
XM_011545790.2 | XP_011544092.1 | 3, 4 | 1236 | |||
GenBank mRNA | AB449895.1 | - | 3, 4 | 696 | ||
AF370370.1 | - | 3, 4 | 324 | |||
AK023192.1 | - | 3, 4 | 558 | |||
BC063112.1 | - | 3, 4 | 603 | |||
BC072453.1 | - | 3, 4 | 491 | 6184429 | ||
BC093814.1 | - | 3, 4 | 407 | 7939659 | ||
BC093816.1 | - | 3, 4 | 407 | 7939661 | ||
BC098419.1 | - | 3, 4 | 743 |