Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320356.1 | NP_001307285.1 | 6, 7 | 889 | ||
NM_001320357.1 | NP_001307286.1 | 5, 6 | 779 | |||
NM_001320358.1 | NP_001307287.1 | 5, 6 | 649 | |||
NM_001320359.1 | NP_001307288.1 | 6, 7 | 805 | |||
NM_001320360.1 | NP_001307289.1 | 5, 6 | 1035 | |||
NM_001320361.1 | NP_001307290.1 | 6, 7 | 1019 | |||
NM_001320362.1 | NP_001307291.1 | 5, 6 | 599 | |||
NM_001320363.1 | NP_001307292.1 | 5, 6 | 729 | |||
NM_001320364.1 | NP_001307293.1 | 4, 5 | 658 | |||
NM_001320365.1 | NP_001307294.1 | 6, 7 | 969 | |||
NM_015528.2 | NP_056343.1 | 5, 6 | 1165 | |||
XM_017024427.1 | XP_016879916.1 | 4, 5 | 905 | |||
GenBank mRNA | AK025329.1 | - | 5, 6 | 838 | ||
AL050060.1 | - | 3, 4 | 315 | |||
AL537131.3 | - | 6, 7 | 693 | |||
AL834284.1 | - | 4, 5 | 687 | |||
AY203930.1 | - | 4, 5 | 665 | |||
AY960626.1 | - | 4, 5 | 332 | |||
BC010139.2 | - | 4, 5 | 563 | 4128232 | ||
BC013222.1 | - | 4, 5 | 619 | |||
BI825561.1 | - | 5, 6 | 501 | |||
CB150714.1 | - | 5, 6 | 551 | |||
CR457340.1 | - | 4, 5 | 389 |