Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015529.3 | NP_056344.2 | 5, 6 | 959 | ||
XM_017010714.1 | XP_016866203.1 | 5, 6 | 767 | |||
XM_017010715.1 | XP_016866204.1 | 5, 6 | 605 | |||
GenBank mRNA | AK074879.1 | - | 5, 6 | 674 | ||
AK226054.1 | - | 1, 2 | 56 | |||
AL080058.1 | - | 17 | ||||
AY007239.1 | - | 4, 5 | 875 | |||
AY359094.1 | - | 5, 6 | 883 | |||
BC018756.1 | - | 5, 6 | 918 | 4841330 |