Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015542.3 | NP_056357.1 | 2 | 323 | ||
NM_080599.2 | NP_542166.1 | 2 | 493 | |||
XM_011519449.2 | XP_011517751.1 | 2 | 371 | |||
GenBank mRNA | AB037829.1 | - | 1 | 692 | ||
AF301013.1 | - | 2 | 316 | |||
AF318574.1 | - | 2 | 293 | |||
AK310925.1 | - | 2 | 282 | |||
AY013249.1 | - | 1 | 242 | |||
BC090919.1 | - | 2 | 291 | |||
BC114964.1 | - | 2 | 287 | 40082739 | ||
BC115737.1 | - | 2 | 287 | 40082728 | ||
DA188353.1 | - | 2 | 323 |