Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SIPA1L1 SIRNA Assays ›
Gene Symbol:
SIPA1L1
Gene Name:
signal induced proliferation associated 1 like 1
Gene Aliases:
E6TP1
Chromosome Location:
Chr.14: 71320418 - 71741229 on Build GRCh38
UniGene ID:
Hs.654657
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
23079
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SIPA1L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284245.1
NP_001271174.1
2
663
NM_001284246.1
NP_001271175.1
2
1252
NM_001284247.1
NP_001271176.1
1
604
NM_015556.2
NP_056371.1
2
663
XM_005267516.4
XP_005267573.1
2
1252
XM_006720111.2
XP_006720174.1
4
2542
XM_011536630.1
XP_011534932.1
6
1014
XM_011536631.1
XP_011534933.1
6
1425
XM_011536632.1
XP_011534934.1
5
934
XM_011536635.2
XP_011534937.1
5
1081
XM_017021159.1
XP_016876648.1
8
1171
XM_017021160.1
XP_016876649.1
6
1093
XM_017021161.1
XP_016876650.1
7
1046
XM_017021162.1
XP_016876651.1
9
1228
XM_017021163.1
XP_016876652.1
4
895
XM_017021164.1
XP_016876653.1
4
902
XM_017021165.1
XP_016876654.1
6
971
XM_017021166.1
XP_016876655.1
7
1104
XM_017021167.1
XP_016876656.1
6
979
XM_017021168.1
XP_016876657.1
6
953
XM_017021169.1
XP_016876658.1
5
1106
XM_017021170.1
XP_016876659.1
5
1464
XM_017021171.1
XP_016876660.1
6
1037
XM_017021172.1
XP_016876661.1
5
912
XM_017021173.1
XP_016876662.1
4
1039
XM_017021174.1
XP_016876663.1
4
1279
XM_017021175.1
XP_016876664.1
4
1397
XM_017021176.1
XP_016876665.1
9
1224
XM_017021177.1
XP_016876666.1
9
1213
XM_017021178.1
XP_016876667.1
7
1051
XM_017021179.1
XP_016876668.1
6
954
XM_017021180.1
XP_016876669.1
10
1266
XM_017021181.1
XP_016876670.1
8
1171
XM_017021182.1
XP_016876671.1
7
1104
XM_017021183.1
XP_016876672.1
5
912
XM_017021184.1
XP_016876673.1
9
1224
XM_017021185.1
XP_016876674.1
2
1252
XM_017021186.1
XP_016876675.1
8
1171
XM_017021187.1
XP_016876676.1
6
1037
XM_017021188.1
XP_016876677.1
5
912
XM_017021189.1
XP_016876678.1
7
1021
XM_017021190.1
XP_016876679.1
8
1171
XM_017021191.1
XP_016876680.1
6
979
XM_017021192.1
XP_016876681.1
5
912
XM_017021193.1
XP_016876682.1
5
910
XM_017021194.1
XP_016876683.1
8
1171
XM_017021195.1
XP_016876684.1
8
1171
XM_017021196.1
XP_016876685.1
8
1171
GenBank mRNA
AB007900.2
-
1
387
AF090989.1
-
2
650
AF090990.1
-
2
650
AK122930.1
-
2
1252
BC146777.1
-
1
387
9007226
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_139330, AY043226, AF026504.This siRNA also targets the following transcripts in Mouse: NM_172579, NM_001167983, BC058681, AK171876, AK029889, and others.
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
actin cytoskeleton reorganization
regulation of GTPase activity
ephrin receptor signaling pathway
regulation of synaptic plasticity
regulation of axonogenesis
regulation of small GTPase mediated signal transduction
regulation of dendritic spine morphogenesis
activation of GTPase activity
molecular_function
GTPase activator activity
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline