Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100409.2 | NP_001093879.1 | 3 | 1204 | ||
NM_001304792.1 | NP_001291721.1 | 3 | 1204 | |||
NM_015571.3 | NP_056386.2 | 3 | 1204 | |||
XM_006715457.3 | XP_006715520.1 | 4 | 957 | |||
XM_011535746.2 | XP_011534048.1 | 2 | 755 | |||
XM_011535747.2 | XP_011534049.1 | 2 | 1110 | |||
XM_017010719.1 | XP_016866208.1 | 3 | 849 | |||
XM_017010720.1 | XP_016866209.1 | 3 | 161 | |||
XM_017010721.1 | XP_016866210.1 | 3 | 169 | |||
XM_017010722.1 | XP_016866211.1 | 4 | 977 | |||
XM_017010723.1 | XP_016866212.1 | 4 | 957 | |||
XM_017010724.1 | XP_016866213.1 | 4 | 1332 | |||
XM_017010725.1 | XP_016866214.1 | 2 | 1110 | |||
GenBank mRNA | AB018340.2 | - | 3 | 232 | ||
AF196304.1 | - | 3 | 188 | |||
AF306508.1 | - | 3 | 203 | |||
AF307849.1 | - | 3 | 263 | |||
AK096455.1 | - | 3 | 786 | |||
AK126921.1 | - | 3 | 1204 | |||
BC028583.1 | - | 3 | 411 | 4838569 |