Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042469.1 | NP_001035934.2 | 8 | 1389 | ||
NM_001042470.1 | NP_001035935.2 | 6 | 1182 | |||
NM_001130069.2 | NP_001123541.1 | 8 | 1289 | |||
NM_001146333.1 | NP_001139805.1 | 8 | 1277 | |||
NM_015411.2 | NP_056226.2 | 9 | 1434 | |||
XM_017011939.1 | XP_016867428.1 | 10 | 1552 | |||
XM_017011940.1 | XP_016867429.1 | 9 | 1724 | |||
GenBank mRNA | AK075477.1 | - | 9 | 1393 | ||
AK301627.1 | - | 6 | 1099 | |||
AL050037.1 | - | 7 | 1153 | |||
AY359103.1 | - | 9 | 1364 | |||
BC000224.1 | - | 8 | 1169 | |||
BC006159.1 | - | 9 | 1332 | |||
BC015600.2 | - | 8 | 1186 | |||
BC065222.1 | - | 6 | 1070 | |||
BC084539.1 | - | 9 | 1364 | 6599080 | ||
CR936749.1 | - | 8 | 1324 | |||
CR936757.1 | - | 9 | 1377 |