Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHCTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323342.1 | NP_001310271.1 | 30 | 4595 | ||
NM_001323343.1 | NP_001310272.1 | 30 | 4595 | |||
NM_015446.4 | NP_056261.4 | 30 | 4311 | |||
NR_136586.1 | 30 | 4595 | ||||
XM_006711758.1 | XP_006711821.1 | 30 | 4424 | |||
XM_011544156.2 | XP_011542458.2 | 30 | 4290 | |||
XM_011544157.2 | XP_011542459.2 | 30 | 4290 | |||
GenBank mRNA | AB059277.1 | - | 30 | 4311 | ||
AK091510.1 | - | 8 | 1370 | |||
AK095398.1 | - | 4 | 848 | |||
AK122844.1 | - | 14 | 2084 | |||
AL832213.1 | - | 21 | 2802 | |||
AL833491.1 | - | 15 | 2195 | |||
AY157619.1 | - | 30 | 4430 |
Gene Symbol: AHCTF1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077058.1 | 1 | 4409 |