Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098616.2 | NP_001092086.1 | 7 | 1718 | ||
NM_001297717.1 | NP_001284646.1 | 5 | 1605 | |||
NM_001297718.1 | NP_001284647.1 | 7 | 1641 | |||
NM_001297720.1 | NP_001284649.1 | 6 | 1664 | |||
NM_015449.3 | NP_056264.1 | 6 | 1616 | |||
NM_138740.3 | NP_620077.1 | 5 | 1562 | |||
GenBank mRNA | AF077036.1 | - | 6 | 1436 | ||
AF109185.1 | - | 7 | 1409 | |||
AJ243663.1 | - | 5 | 1384 | |||
AJ243664.1 | - | 6 | 1384 | |||
AJ243665.1 | - | 6 | 1221 | |||
AL117463.1 | - | 5 | 1182 | |||
BC000152.2 | - | 7 | 1527 | 2901006 | ||
BC001377.2 | - | 6 | 1384 | 3051048 | ||
BC006514.1 | - | 6 | 1453 | 2822327 | ||
BC014391.1 | - | 6 | 1352 | 3544590 | ||
BC088353.1 | - | 5 | 1425 | 6164281 | ||
BU072807.1 | - | 1 | 411 | |||
CF123260.1 | - | 224 |