Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015470.2 | NP_056285.1 | 5 | 4281 | ||
XM_005264251.3 | XP_005264308.1 | 6 | 5917 | |||
XM_005264252.3 | XP_005264309.1 | 6 | 6235 | |||
XM_006711985.3 | XP_006712048.1 | 7 | 6010 | |||
XM_011532753.2 | XP_011531055.1 | 7 | 5520 | |||
XM_017003788.1 | XP_016859277.1 | 7 | 5572 | |||
GenBank mRNA | AB020664.1 | - | 5 | 4281 | ||
AF334812.1 | - | 5 | 4280 | |||
AK023823.1 | - | 4 | 3946 | |||
AK024727.1 | - | 1 | 1733 | |||
AL117579.1 | - | 3 | 2841 | |||
BC035013.1 | - | 5 | 4204 | 5269818 |