Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015509.3 | NP_056324.2 | 8 | 948 | ||
NR_024260.1 | 7 | 866 | ||||
GenBank mRNA | AK074858.1 | - | 8 | 905 | ||
AK074880.1 | - | 8 | 918 | |||
AK074923.1 | - | 8 | 891 | |||
AK075013.1 | - | 8 | 925 | |||
AK094805.1 | - | 3 | 1490 | |||
AK290537.1 | - | 7 | 791 | |||
AK303675.1 | - | 6 | 474 | |||
AL050272.1 | - | 7 | 792 | |||
BC002888.2 | - | 8 | 850 | |||
BC067367.1 | - | 6 | 620 | |||
BC084551.1 | - | 4 | 459 | |||
BC110876.1 | - | 8 | 882 | 5553942 |