Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199570.1 | NP_001186499.1 | 13 | 2933 | ||
NM_015526.2 | NP_056341.1 | 14 | 2849 | |||
GenBank mRNA | AJ427922.1 | - | 14 | 2827 | ||
AK056445.1 | - | 12 | 2332 | |||
AK092354.1 | - | 10 | 2096 | |||
AK094738.1 | - | 14 | 2847 | |||
AL117468.1 | - | 6 | 1461 | |||
BC014486.1 | - | 11 | 2292 |