Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015527.3 | NP_056342.3 | 9 | 3264 | ||
XM_011545789.1 | XP_011544091.1 | 6 | 3006 | |||
XM_011545790.2 | XP_011544092.1 | 8 | 3131 | |||
GenBank mRNA | AF370370.1 | - | 9 | 2411 | ||
AK002101.1 | - | 3 | 1767 | |||
AK023192.1 | - | 9 | 2643 | |||
AL117408.1 | - | 2 | 2461 | |||
AY007150.1 | - | 1 | 983 | |||
BC050523.2 | - | 5 | 1978 | |||
BC063112.1 | - | 9 | 2688 | |||
BC072453.1 | - | 9 | 2576 | 6184429 | ||
BC098419.1 | - | 9 | 2828 |