Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114093.1 | NP_001107565.1 | 11 | 1841 | ||
NM_015578.2 | NP_056393.2 | 10 | 1784 | |||
XM_017026575.1 | XP_016882064.1 | 9 | 1626 | |||
XM_017026576.1 | XP_016882065.1 | 10 | 1683 | |||
XM_017026577.1 | XP_016882066.1 | 9 | 1620 | |||
GenBank mRNA | AB069974.1 | - | 11 | 1720 | ||
AB069975.1 | - | 10 | 1663 | |||
AK027369.1 | - | 9 | 1394 | |||
AK027643.1 | - | 10 | 1665 | |||
AK127401.1 | - | 7 | 1907 | |||
AK300208.1 | - | 10 | 1640 | |||
AL117499.1 | - | 5 | 725 | |||
AL833221.1 | - | 8 | 1257 | |||
AL834398.1 | - | 11 | 1750 | |||
BC016842.1 | - | 10 | 1784 | 3902704 |