Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202438.1 | NP_001189367.1 | 17, 18 | 2672 | ||
NM_015608.2 | NP_056423.2 | 16, 17 | 2570 | |||
NR_110857.1 | 17, 18 | 2667 | ||||
NR_110858.1 | 18, 19 | 2798 | ||||
NR_110859.1 | 17, 18 | 2688 | ||||
XM_011539622.1 | XP_011537924.1 | 14, 15 | 1860 | |||
XM_017016066.1 | XP_016871555.1 | 17, 18 | 2416 | |||
GenBank mRNA | AK091507.1 | - | 16, 17 | 2382 | ||
AK314958.1 | - | 2, 3 | 406 | |||
AL050102.1 | - | 1, 2 | 148 | |||
BC016903.1 | - | 5, 6 | 668 | |||
BC026172.1 | - | 18, 19 | 2799 | 4838438 | ||
BC105929.1 | - | 17, 18 | 2606 | |||
BC107479.1 | - | 17, 18 | 2611 | 40028896 |