Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015621.2 | NP_056436.2 | 9 | 1073 | ||
GenBank mRNA | AK023767.1 | - | 8 | 851 | ||
AK094658.1 | - | 2 | 1109 | |||
AK225826.1 | - | 9 | 1043 | |||
AK292841.1 | - | 9 | 1073 | |||
AK298063.1 | - | 4 | 343 | |||
AL080169.1 | - | 2 | 273 | |||
AL713742.1 | - | 8 | 997 | |||
AY538626.1 | - | 2 | 283 | |||
BC016647.1 | - | 8 | 886 | |||
BM927471.1 | - | 4 | 590 |