Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015630.3 | NP_056445.3 | 2 | 309 | ||
XM_011510941.2 | XP_011509243.1 | 2 | 520 | |||
GenBank mRNA | AF286904.1 | - | 2 | 301 | ||
AK125789.1 | - | 3 | 378 | |||
BC093818.1 | - | 2 | 221 | 7939663 | ||
BC093820.1 | - | 2 | 221 | 7939665 |