Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278431.1 | NP_001265360.1 | 3 | 1120 | ||
NM_015645.4 | NP_056460.1 | 15 | 3684 | |||
GenBank mRNA | AF086482.1 | - | 1 | 198 | ||
AF329841.1 | - | 3 | 1085 | |||
AL110261.1 | - | 2 | 816 | |||
AY358383.1 | - | 3 | 1114 | |||
BC029485.1 | - | 3 | 1121 |
Gene Symbol: MFRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031433.3 | NP_113621.1 | 15 | 3684 | ||
GenBank mRNA | AJ862823.1 | - | 15 | 3666 |