Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015705.5 | NP_056520.2 | 3 | 312 | ||
XM_005261572.2 | XP_005261629.1 | 3 | 339 | |||
XM_005261573.3 | XP_005261630.1 | 3 | 344 | |||
XM_005261574.4 | XP_005261631.1 | 3 | 344 | |||
XM_011530158.1 | XP_011528460.1 | 3 | 307 | |||
XM_011530159.1 | XP_011528461.1 | 3 | 289 | |||
XM_011530161.1 | XP_011528463.1 | 3 | 289 | |||
XM_011530162.2 | XP_011528464.1 | 3 | 284 | |||
GenBank mRNA | AB196956.1 | - | 1 | 81 | ||
AB275763.1 | - | 2 | 185 | |||
AB449880.1 | - | 1 | 74 | |||
AK295469.1 | - | 3 | 258 | |||
AK309400.1 | - | 3 | 268 | |||
AY308849.1 | - | 3 | 247 | |||
BC008078.2 | - | 3 | 238 | 3027777 | ||
BP283990.1 | - | 3 | 312 | |||
BU585020.1 | - | 3 | 287 | |||
CR456441.1 | - | 2 | 110 |