Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015717.4 | NP_056532.4 | 3 | 302 | ||
XM_011532874.1 | XP_011531176.1 | 4 | 490 | |||
XM_011532875.2 | XP_011531177.1 | 5 | 507 | |||
XM_011532876.2 | XP_011531178.1 | 5 | 506 | |||
GenBank mRNA | AJ242859.1 | - | 3 | 302 | ||
AK314927.1 | - | 3 | 296 | |||
BC022278.1 | - | 3 | 295 | 4692155 |