Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015720.3 | NP_056535.1 | 3 | 1178 | ||
GenBank mRNA | AF219137.1 | - | 3 | 1137 | ||
AY359096.1 | - | 3 | 1141 | |||
BC052585.1 | - | 3 | 1133 | 6264322 |