Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF691 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242739.1 | NP_001229668.1 | 4 | 630 | ||
NM_015911.3 | NP_056995.1 | 2 | 329 | |||
XM_006710671.3 | XP_006710734.1 | 3 | 446 | |||
XM_017001400.1 | XP_016856889.1 | 3 | 775 | |||
XM_017001401.1 | XP_016856890.1 | 2 | 3312 | |||
XM_017001402.1 | XP_016856891.1 | 3 | 1776 | |||
XM_017001403.1 | XP_016856892.1 | 2 | 4312 | |||
GenBank mRNA | AF131739.1 | - | 2 | 293 | ||
AK000938.1 | - | 2 | 432 | |||
AK296201.1 | - | 3 | 718 | |||
BC000157.2 | - | 2 | 372 | 2901198 | ||
BM741761.1 | - | 3 | 443 | |||
CR457146.1 | - | 1 | 235 |