Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202438.1 | NP_001189367.1 | 18 | 2743 | ||
NM_015608.2 | NP_056423.2 | 17 | 2641 | |||
NR_110857.1 | 18 | 2738 | ||||
NR_110858.1 | 19 | 2869 | ||||
NR_110859.1 | 18 | 2759 | ||||
XM_011539622.1 | XP_011537924.1 | 15 | 1931 | |||
XM_017016066.1 | XP_016871555.1 | 18 | 2487 | |||
GenBank mRNA | AK091507.1 | - | 17 | 2453 | ||
AK314958.1 | - | 3 | 477 | |||
AL050102.1 | - | 2 | 219 | |||
BC016903.1 | - | 6 | 739 | |||
BC026172.1 | - | 19 | 2870 | 4838438 | ||
BC105929.1 | - | 18 | 2677 | |||
BC107479.1 | - | 18 | 2682 | 40028896 |