Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015613.2 | NP_056428.1 | 4 | 1386 | ||
XM_011539623.2 | XP_011537925.1 | 3 | 1642 | |||
XM_011539626.2 | XP_011537928.1 | 3 | 937 | |||
GenBank mRNA | AB031547.1 | - | 4 | 1386 | ||
AL080175.1 | - | 1 | 459 |