Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005502.3 | NP_005493.2 | 4 | 605 | ||
XM_005251773.2 | XP_005251830.1 | 4 | 514 | |||
XM_005251776.2 | XP_005251833.1 | 3 | 354 | |||
XM_011518339.2 | XP_011516641.1 | 5 | 589 | |||
XM_011518340.2 | XP_011516642.1 | 6 | 804 | |||
XM_011518341.2 | XP_011516643.1 | 5 | 589 | |||
XM_011518344.2 | XP_011516646.1 | 5 | 589 | |||
XM_017014378.1 | XP_016869867.1 | 5 | 787 | |||
XM_017014379.1 | XP_016869868.1 | 5 | 1195 | |||
XM_017014380.1 | XP_016869869.1 | 5 | 521 | |||
XM_017014381.1 | XP_016869870.1 | 5 | 587 | |||
XM_017014382.1 | XP_016869871.1 | 3 | 1052 | |||
GenBank mRNA | AB037924.1 | - | 3 | 288 | ||
AB055982.1 | - | 3 | 201 | |||
AB445477.1 | - | 3 | 201 | |||
AF165281.1 | - | 2 | 141 | |||
AF258627.1 | - | 4 | 596 | |||
AF285167.1 | - | 4 | 491 | |||
AJ012376.1 | - | 2 | 141 | |||
AK024328.1 | - | 4 | 514 | |||
BC146856.1 | - | 4 | 415 | 9056784 | ||
BX955241.1 | - | 4 | 605 |