Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3YL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159597.2 | NP_001153069.1 | 9 | 1149 | ||
NM_001282682.1 | NP_001269611.1 | 11 | 1254 | |||
NM_001282687.1 | NP_001269616.1 | 12 | 1360 | |||
NM_015677.3 | NP_056492.2 | 10 | 1206 | |||
NR_104223.1 | 12 | 2765 | ||||
NR_104225.1 | 10 | 1397 | ||||
NR_104226.1 | 11 | 1869 | ||||
NR_104227.1 | 11 | 2691 | ||||
GenBank mRNA | AK026507.1 | - | 9 | 1084 | ||
AK026586.1 | - | 10 | 1397 | |||
AK123829.1 | - | 12 | 2947 | |||
AK292312.1 | - | 11 | 1198 | |||
AL050373.1 | - | 12 | 1373 | |||
BC008374.1 | - | 10 | 1115 | 3996066 | ||
BC008375.1 | - | 9 | 1076 | 3996120 |