Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002755.2 | NP_001002755.1 | 4 | 536 | ||
NM_001002756.2 | NP_001002756.1 | 3 | 490 | |||
NM_015700.3 | NP_056515.2 | 4 | 626 | |||
NR_045631.1 | 3 | 400 | ||||
NR_045632.1 | 3 | 400 | ||||
XM_017003808.1 | XP_016859297.1 | 4 | 339 | |||
GenBank mRNA | AF132967.1 | - | 4 | 335 | ||
AJ132584.1 | - | 4 | 414 | |||
AK300700.1 | - | 4 | 345 | |||
AK308567.1 | - | 3 | 207 | |||
AK314004.1 | - | 4 | 369 | |||
AY286306.1 | - | 4 | 383 | |||
AY286307.1 | - | 4 | 473 | |||
AY335194.1 | - | 4 | 529 | |||
BC113692.1 | - | 4 | 498 | 8322744 | ||
BC113694.1 | - | 4 | 498 | 8322746 | ||
BG385940.1 | - | 3 | 288 | |||
BM846341.1 | - | 4 | 536 | |||
BU178674.1 | - | 3 | 232 | |||
BU507991.1 | - | 4 | 341 | |||
BX538347.1 | - | 4 | 401 | |||
DB496651.2 | - | 4 | 522 |