Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLTSCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015711.3 | NP_056526.3 | 10 | 3310 | ||
XM_005258833.4 | XP_005258890.1 | 10 | 3353 | |||
XM_011526882.2 | XP_011525184.1 | 6 | 4320 | |||
XM_011526883.2 | XP_011525185.1 | 9 | 3176 | |||
GenBank mRNA | AF182077.1 | - | 5 | 3326 |