Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VILL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015873.3 | NP_056957.3 | 7, 8 | 931 | ||
XM_005265191.3 | XP_005265248.1 | 8, 9 | 1039 | |||
XM_005265192.4 | XP_005265249.1 | 8, 9 | 2174 | |||
XM_006713184.3 | XP_006713247.1 | 9, 10 | 1039 | |||
XM_011533770.2 | XP_011532072.1 | 8, 9 | 1195 | |||
XM_011533771.1 | XP_011532073.1 | 8, 9 | 1195 | |||
XM_011533773.2 | XP_011532075.1 | 8, 9 | 1196 | |||
GenBank mRNA | AK309304.1 | - | 5, 6 | 818 | ||
BC000243.1 | - | 4, 5 | 467 | 3356294 | ||
BC004300.1 | - | 961 | 3622951 | |||
D88154.1 | - | 3, 4 | 337 | |||
KU178619.1 | - | 3, 4 | 335 | |||
KU178620.1 | - | 3, 4 | 335 |