Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015603.2 | NP_056418.1 | 6 | 697 | ||
XM_017026578.1 | XP_016882067.1 | 6 | 742 | |||
GenBank mRNA | AK302153.1 | - | 7 | 618 | ||
AL050284.1 | - | 6 | 636 | |||
BC002787.2 | - | 6 | 593 | 3615928 | ||
BC009743.2 | - | 6 | 593 | 3832697 |