Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165937.1 | NP_001159409.1 | 3 | 440 | ||
NM_001165938.1 | NP_001159410.1 | 3 | 440 | |||
NM_006804.3 | NP_006795.3 | 3 | 440 | |||
XM_006721646.2 | XP_006721709.1 | 3 | 409 | |||
XM_011524210.1 | XP_011522512.1 | 3 | 409 | |||
XM_011524211.2 | XP_011522513.1 | 3 | 412 | |||
XM_011524213.1 | XP_011522515.1 | 3 | 409 | |||
XM_017024041.1 | XP_016879530.1 | 3 | 412 | |||
GenBank mRNA | AK225069.1 | - | 3 | 373 | ||
AK300176.1 | - | 3 | 376 | |||
AK300842.1 | - | 3 | 330 | |||
AK301524.1 | - | 3 | 378 | |||
AK303114.1 | - | 3 | 378 | |||
BC008356.1 | - | 3 | 391 | 3547271 | ||
BC008747.2 | - | 3 | 360 | 3346043 | ||
BC025679.1 | - | 3 | 361 | 5203654 | ||
BT006964.1 | - | 2 | 237 | |||
D38255.1 | - | 3 | 358 | |||
DC367486.1 | - | 3 | 440 | |||
X80198.1 | - | 3 | 346 |