Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF2BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015649.2 | NP_056464.1 | 1 | 2490 | ||
GenBank mRNA | AK092407.1 | - | 1 | 1745 | ||
AL080159.1 | - | 1 | 1427 | |||
AY278022.1 | - | 1 | 2441 | |||
BC005834.2 | - | 1 | 1878 | |||
BC038222.1 | - | 1 | 2438 | 5168442 | ||
BC039002.1 | - | 1 | 2455 | 5182566 | ||
BC078181.1 | - | 1 | 2163 | 6501537 |