Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290261.1 | NP_001277190.1 | 4 | 2198 | ||
NM_015655.3 | NP_056470.1 | 5 | 1871 | |||
XM_005260702.3 | XP_005260759.1 | 4 | 1687 | |||
XM_006723558.3 | XP_006723621.1 | 5 | 2874 | |||
XM_011529219.2 | XP_011527521.1 | 5 | 1994 | |||
XM_017027802.1 | XP_016883291.1 | 3 | 3769 | |||
XM_017027803.1 | XP_016883292.1 | 4 | 2778 | |||
XM_017027804.1 | XP_016883293.1 | 2 | 1547 | |||
GenBank mRNA | AK056184.1 | - | 5 | 1782 | ||
AK056466.1 | - | 1 | 1394 | |||
AK291235.1 | - | 5 | 1808 | |||
AK299811.1 | - | 3 | 1873 | |||
AL049942.1 | - | 1 | 912 | |||
BC021298.2 | - | 5 | 1800 | 4551314 |