Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321389.1 | NP_001308318.1 | 9 | 2333 | ||
NM_001321390.1 | NP_001308319.1 | 9 | 1978 | |||
NM_001321391.1 | NP_001308320.1 | 9 | 2012 | |||
NM_015680.5 | NP_056495.3 | 8 | 2070 | |||
NR_135628.1 | 8 | 1950 | ||||
NR_135629.1 | 7 | 2008 | ||||
GenBank mRNA | AF070638.1 | - | 2 | 1332 | ||
AK075274.1 | - | 8 | 2026 | |||
AY007160.1 | - | 8 | 2043 | |||
BC001393.2 | - | 8 | 1993 | 3051397 | ||
BC012821.2 | - | 8 | 2042 | 2958621 |