Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002755.2 | NP_001002755.1 | 5, 6 | 698 | ||
NM_001002756.2 | NP_001002756.1 | 4, 5 | 652 | |||
NM_015700.3 | NP_056515.2 | 5, 6 | 788 | |||
NR_045631.1 | 4, 5 | 562 | ||||
NR_045632.1 | 4, 5 | 562 | ||||
XM_017003808.1 | XP_016859297.1 | 5, 6 | 501 | |||
GenBank mRNA | AF132967.1 | - | 5, 6 | 500 | ||
AJ132584.1 | - | 5, 6 | 576 | |||
AK300700.1 | - | 5, 6 | 507 | |||
AK308567.1 | - | 4, 5 | 369 | |||
AK314004.1 | - | 5, 6 | 531 | |||
AY286306.1 | - | 5, 6 | 545 | |||
AY286307.1 | - | 5, 6 | 635 | |||
AY335194.1 | - | 5, 6 | 691 | |||
BC113692.1 | - | 5, 6 | 660 | 8322744 | ||
BC113694.1 | - | 5, 6 | 660 | 8322746 | ||
BG385940.1 | - | 4, 5 | 450 | |||
BU178674.1 | - | 4, 5 | 394 | |||
BU507991.1 | - | 5, 6 | 503 | |||
BX538347.1 | - | 5, 6 | 563 |