Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015705.5 | NP_056520.2 | 4, 5 | 399 | ||
XM_005261572.2 | XP_005261629.1 | 4, 5 | 426 | |||
XM_005261573.3 | XP_005261630.1 | 4, 5 | 431 | |||
XM_005261574.4 | XP_005261631.1 | 4, 5 | 431 | |||
XM_005261575.1 | XP_005261632.1 | 3, 4 | 294 | |||
XM_011530158.1 | XP_011528460.1 | 4, 5 | 394 | |||
XM_011530161.1 | XP_011528463.1 | 4, 5 | 376 | |||
XM_017028776.1 | XP_016884265.1 | 3, 4 | 287 | |||
XM_017028777.1 | XP_016884266.1 | 3, 4 | 287 | |||
XM_017028778.1 | XP_016884267.1 | 3, 4 | 294 | |||
XM_017028779.1 | XP_016884268.1 | 3, 4 | 293 | |||
XM_017028780.1 | XP_016884269.1 | 3, 4 | 282 | |||
GenBank mRNA | AB196956.1 | - | 2, 3 | 168 | ||
AB275763.1 | - | 3, 4 | 272 | |||
AB449880.1 | - | 2, 3 | 161 | |||
AK295469.1 | - | 4, 5 | 345 | |||
AK297599.1 | - | 3, 4 | 247 | |||
AK309400.1 | - | 4, 5 | 355 | |||
AY308849.1 | - | 4, 5 | 334 | |||
BC008078.2 | - | 4, 5 | 325 | 3027777 | ||
BP283990.1 | - | 4, 5 | 399 | |||
BU585020.1 | - | 4, 5 | 374 | |||
CR456441.1 | - | 3, 4 | 197 |