Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WT1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC002734.2 | - | 1 | 1051 | ||
DQ289490.1 | - | 2 | 2030 | |||
M60614.1 | - | 1 | 1898 |