Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VILL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015873.3 | NP_056957.3 | 9, 10 | 1173 | ||
XM_005265191.3 | XP_005265248.1 | 10, 11 | 1281 | |||
XM_005265192.4 | XP_005265249.1 | 10, 11 | 2416 | |||
XM_006713184.3 | XP_006713247.1 | 11, 12 | 1281 | |||
XM_011533770.2 | XP_011532072.1 | 10, 11 | 1437 | |||
XM_011533771.1 | XP_011532073.1 | 10, 11 | 1437 | |||
XM_011533773.2 | XP_011532075.1 | 10, 11 | 1438 | |||
GenBank mRNA | AK309304.1 | - | 7, 8 | 1063 | ||
BC000243.1 | - | 6, 7 | 709 | 3356294 | ||
BC004300.1 | - | 10, 11 | 1203 | 3622951 | ||
D88154.1 | - | 5, 6 | 579 | |||
KU178619.1 | - | 5, 6 | 577 | |||
KU178620.1 | - | 5, 6 | 577 |