Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015909.3 | NP_056993.2 | 38 | 4610 | ||
NR_052013.2 | 38 | 4610 | ||||
XM_011510357.2 | XP_011508659.1 | 37 | 4477 | |||
XM_011510358.2 | XP_011508660.1 | 38 | 4606 | |||
XM_011510359.2 | XP_011508661.1 | 32 | 4011 | |||
XM_011510360.2 | XP_011508662.1 | 18 | 2430 | |||
XM_011510361.2 | XP_011508663.1 | 18 | 2421 | |||
XM_017004317.1 | XP_016859806.1 | 38 | 4606 | |||
GenBank mRNA | AB621813.1 | - | 38 | 4610 | ||
AF056195.1 | - | 17 | 1812 | |||
AF388385.1 | - | 38 | 4592 | |||
AL050281.1 | - | 11 | 1379 | |||
AL832774.1 | - | 38 | 4553 | |||
AL833270.1 | - | 38 | 4570 | |||
BC040345.1 | - | 30 | 3878 | |||
BC051792.1 | - | 14 | 1709 | |||
BC131735.1 | - | 38 | 4592 | 40125600 | ||
HM243530.1 | - | 2 | 156 | |||
HM243532.1 | - | 3 | 351 |