Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000078.2 | NP_000069.2 | 9 | 941 | ||
GenBank mRNA | AK291586.1 | - | 9 | 901 | ||
BC025739.1 | - | 9 | 900 | 5223614 | ||
KU177972.1 | - | 9 | 884 | |||
M30185.1 | - | 9 | 1014 |