Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014433.2 | NP_001014433.1 | 4 | 499 | ||
NM_001014837.1 | NP_001014837.1 | 4 | 748 | |||
NM_001014838.1 | NP_001014838.1 | 4 | 717 | |||
NM_001014840.1 | NP_001014840.1 | 4 | 627 | |||
NM_015921.2 | NP_057005.1 | 3 | 890 | |||
GenBank mRNA | AF106943.1 | - | 3 | 527 | ||
AF230924.1 | - | 3 | 313 | |||
AK311556.1 | - | 1 | 987 | |||
AL533788.3 | - | 2 | 818 | |||
BC005890.2 | - | 4 | 324 | 3505574 | ||
BC107751.1 | - | 3 | 527 | 6714433 | ||
BU540844.1 | - | 505 | ||||
CB106834.1 | - | 4 | 453 | |||
CN363642.1 | - | 4 | 529 |