Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129884.2 | NP_001123356.1 | 5 | 730 | ||
NM_001243725.1 | NP_001230654.1 | 4 | 838 | |||
NM_001243726.1 | NP_001230655.1 | 4 | 622 | |||
NM_001243727.2 | NP_001230656.1 | 3 | 456 | |||
NM_001278195.1 | NP_001265124.1 | 5 | 770 | |||
NM_015926.5 | NP_057010.1 | 4 | 678 | |||
NR_024012.2 | 3 | 386 | ||||
NR_103462.1 | 3 | 349 | ||||
NR_103463.1 | 3 | 318 | ||||
XM_006713195.3 | XP_006713258.1 | 4 | 618 | |||
XM_006713197.3 | XP_006713260.1 | 4 | 585 | |||
XM_006713198.2 | XP_006713261.1 | 4 | 653 | |||
XM_011533805.2 | XP_011532107.1 | 4 | 1333 | |||
XM_017006573.1 | XP_016862062.1 | 4 | 838 | |||
GenBank mRNA | AF072733.1 | - | 3 | 576 | ||
AK023997.1 | - | 4 | 621 | |||
AK222938.1 | - | 4 | 590 | |||
AK222949.1 | - | 4 | 590 | |||
AY358972.1 | - | 3 | 577 | |||
BC001592.1 | - | 4 | 600 | |||
BC008742.2 | - | 4 | 838 | 3346733 | ||
BG326586.1 | - | 3 | 373 | |||
BG772477.1 | - | 3 | 391 | |||
CB142778.1 | - | 3 | 350 | |||
HY149215.1 | - | 3 | 372 |