Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016024.2 | NP_057108.2 | 2, 3 | 196 | ||
GenBank mRNA | AF078865.1 | - | 1, 2 | 132 | ||
AF151837.1 | - | 1, 2 | 132 | |||
AK292855.1 | - | 2, 3 | 178 | |||
AL050405.1 | - | 2, 3 | 171 | |||
BC003676.1 | - | 2, 3 | 196 | |||
BC033750.1 | - | 2, 3 | 164 | 5210341 | ||
BC040433.1 | - | 2, 3 | 147 | |||
BC105582.1 | - | 2, 3 | 183 |