Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321102.1 | NP_001308031.1 | 2 | 273 | ||
NM_016030.5 | NP_057114.5 | 2 | 310 | |||
XM_011510350.2 | XP_011508652.1 | 2 | 740 | |||
XM_011510352.2 | XP_011508654.1 | 2 | 740 | |||
XM_011510353.2 | XP_011508655.1 | 2 | 740 | |||
XM_011510354.2 | XP_011508656.1 | 2 | 740 | |||
XM_017004257.1 | XP_016859746.1 | 2 | 740 | |||
GenBank mRNA | AK094181.1 | - | 3 | 260 | ||
AK098327.1 | - | 2 | 275 | |||
BC014164.2 | - | 2 | 187 | 4548681 | ||
DA031172.1 | - | 3 | 260 |